Learn More
ACROBiosystems Human Neuropilin-1 / NRP1 / CD304 Protein, Mouse IgG2a Fc Tag (MALS verified)
Brand: ACROBiosystems NR1-H5253-100ug
Additional Details : Weight : 0.00010kg
Description
Neuropilin-1 (NRP1) is also known as Vascular endothelial cell growth factor 165 receptor (VEGF165R), CD antigen CD304, which belongs to the neuropilin family. The membrane-bound isoform 1 is a receptor involved in the development of the cardiovascular system, in angiogenesis, in the formation of certain neuronal circuits and in organogenesis outside the nervous system. It mediates the chemorepulsant activity of semaphorins. It binds to semaphorin 3A, The PLGF-2 isoform of PGF, The VEGF-165 isoform of VEGF and VEGF-B. Coexpression with KDR results in increased VEGF-165 binding to KDR as well as increased chemotaxis. It may regulate VEGF-induced angiogenesis. The soluble isoform 2 binds VEGF-165 and appears to inhibit its binding to cells.
This protein carries a mouse IgG2a Fc tag at the C-terminus. The protein has a calculated MW of 96.7 kDa. The protein migrates as 110-130 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.Specifications
AAH07533.1 | |
100 μg | |
Phe 22 - Lys 644 | |
NRP1,Neuropilin-1,NRP,VEGF165R,CD304 | |
Unconjugated | |
Mouse IgG Fc Tag | |
Less than 0.1 EU / μg by the LAL method. | |
Powder | |
>95% as determined by SDS-PAGE. |
96.7 kDa | |
Human Neuropilin-1, Mouse IgG2a Fc Tag (NR1-H5253) is expressed from human 293 cells (HEK293). It contains AA Phe 22 - Lys 644 (Accession # AAH07533.1). | |
-20°C | |
Neuropilin-1 | |
Human | |
50 mM Tris, 100 mM Glycine, 25 mM Arginine, 150 mM NaCl, pH7.5 | |
HEK293 | |
Bio-Markers & CD Antigens |