missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human NDN Full-length ORF (NP_002478.1, 1 a.a. - 321 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00004692-P01.10ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16100568

  • 335.00€ / 10µg

Estimated Shipment: 12-05-2023
to see stock.
Add to basket

Description

Description

This intronless gene is located in the Prader-Willi syndrome deletion region. It is an imprinted gene and is expressed exclusively from the paternal allele. Studies in mouse suggest that the protein encoded by this gene may suppress growth in postmitotic neurons. [provided by RefSeq]

Sequence: MSEQSKDLSDPNFAAEAPNSEVHSSPGVSEGVPPSATLAEPQSPPLGPTAAPQAAPPPQAPNDEGDPKALQQAAEEGRAHQAPSAAQPGPAPPAPAQLVQKAHELMWYVLVKDQKKMIIWFPDMVKDVIGSYKKWCRSILRRTSLILARVFGLHLRLTSLHTMEFALVKALEPEELDRVALSNRMPMTGLLLMILSLIYVKGRGARESAVWNVLRILGLRPWKKHSTFGDVRKLITEEFVQMNYLKYQRVPYVEPPEYEFFWGSRASREITKMQIMEFLARVFKKDPQAWPSRYREALEEARALREANPTAHYPRSSVSED
Specifications

Specifications

NP_002478.1
Liquid
4692
NDN (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MSEQSKDLSDPNFAAEAPNSEVHSSPGVSEGVPPSATLAEPQSPPLGPTAAPQAAPPPQAPNDEGDPKALQQAAEEGRAHQAPSAAQPGPAPPAPAQLVQKAHELMWYVLVKDQKKMIIWFPDMVKDVIGSYKKWCRSILRRTSLILARVFGLHLRLTSLHTMEFALVKALEPEELDRVALSNRMPMTGLLLMILSLIYVKGRGARESAVWNVLRILGLRPWKKHSTFGDVRKLITEEFVQMNYLKYQRVPYVEPPEYEFFWGSRASREITKMQIMEFLARVFKKDPQAWPSRYREALEEARALREANPTAHYPRSSVSED
RUO
NDN
Wheat Germ (in vitro)
GST
Antibody Production, Array, Enzyme-linked Immunoabsorbent Assay, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
62.5kDa
Glutathione Sepharose 4 Fast Flow
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
HsT16328/PWCR
NDN
Yes
wheat germ expression system
Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers