missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human MSPR/Ron PE-conjugated Antibody, R&D Systems™

Goat Polyclonal Antibody
Brand: R&D Systems FAB6647P
This item is not returnable.
View return policy
Description
MSPR/Ron Polyclonal specifically detects MSPR/Ron in Human samples. It is validated for Flow Cytometry.
Specifications
| MSPR/Ron | |
| Polyclonal | |
| Flow Cytometry 10 uL/10^6 cells | |
| CD136, CD136 antigen, CDw136c-met-related tyrosine kinase, EC 2.7.10, EC 2.7.10.1, macrophage stimulating 1 receptor (c-met-related tyrosine kinase), MSP R, MSP receptor, MST1R, p185-Ron, Protein-tyrosine kinase 8, PTK8, PTK8 protein tyrosine kinase 8, Ron, RONmacrophage-stimulating protein receptor, soluble RON variant 1, soluble RON variant 2, soluble RON variant 3, soluble RON variant 4 | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 4486 | |
| Human | |
| IgG |
| Flow Cytometry | |
| R-PE | |
| Q04912 | |
| MST1R | |
| Mouse myeloma cell line NS0-derived recombinant human MSP R/Ron Glu25-Ser956 Accession # Q04912 | |
| 100 Tests | |
| Primary | |
| Delects human MSP R/Ron in direct ELISAs and Western blots. In these formats, approximately 25% cross-reactivity with recombinant mouse MSP R is observed. | |
| Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction