missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human/Mouse RIPK4/RIP4 Antibody, R&D Systems™

Goat Polyclonal Antibody
Brand: R&D Systems AF10002-SP
This item is not returnable.
View return policy
Description
RIPK4 Polyclonal antibody specifically detects RIPK4 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry
Specifications
| RIPK4 | |
| Polyclonal | |
| Western Blot 2 ug/mL, Immunohistochemistry 0.3-15 ug/mL | |
| P57078 | |
| Goat | |
| Antigen Affinity-purified | |
| RUO | |
| 54101 | |
| Human, Mouse | |
| Purified |
| Western Blot, Immunohistochemistry | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
| ANKK2, ANKRD3, ANKRD3MGC129993, ankyrin repeat domain 3, Ankyrin repeat domain-containing protein 3, DIKMGC129992, EC 2.7.11, EC 2.7.11.1, PKC-delta-interacting protein kinase, PKK, receptor-interacting serine/threonine-protein kinase 4, receptor-interacting serine-threonine kinase 4, RIP4, serine/threonine-protein kinase ANKRD3 | |
| E. coli-derived recombinant human/mouse RIPK4/RIP4, Thr218-Thr450, Accession # P57078 | |
| 25 μg | |
| Primary | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20 to -70 °C under sterile conditions after reconstitution. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction