missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human/Mouse/Rat RAGE/AGER Antibody, R&D Systems™

Goat Polyclonal Antibody has been used in 17 publications
Brand: R&D Systems AF1145
This item is not returnable.
View return policy
Description
AGER Polyclonal specifically detects AGER in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Blockade of Receptor-ligand Interaction.
Specifications
| RAGE | |
| Polyclonal | |
| Unconjugated | |
| Q15109 | |
| AGER | |
| Mouse myeloma cell line NS0-derived recombinant human RAGE Gln24-Ala344 Accession # Q15109 | |
| 100 μg | |
| Primary | |
| Detects human RAGE in direct ELISAs and Western blots. | |
| Human, Mouse, Rat | |
| IgG |
| Western Blot, Immunohistochemistry, Blockade of Receptor-ligand Interaction | |
| LYOPH | |
| Western Blot 1 ug/mL, Immunohistochemistry 0.5-15 ug/mL, Blockade of Receptor-ligand Interaction | |
| advanced glycosylation end product-specific receptor, AGER, RAGE isoform delta, RAGE isoform sRAGE-delta, Receptor for advanced glycosylation end products, receptor for advanced glycosylation end-products, SCARJ1 | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 5891 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction