missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human/Mouse/Rat PP2C alpha/PPM1A Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems MAB4150-SP
This item is not returnable.
View return policy
Description
PP2C alpha/PPM1A Monoclonal specifically detects PP2C alpha/PPM1A in Human, Mouse, Rat samples. It is validated for Western Blot.
Specifications
| PP2C alpha/PPM1A | |
| Monoclonal | |
| LYOPH | |
| Western Blot 1 ug/mL | |
| P35813 | |
| PPM1A | |
| E. coli-derived recombinant human PP2C alpha/PPM1A Met1-Gly324 Accession # P35813 | |
| 25 μg | |
| Primary | |
| Detects human, mouse, and rat PP2C alpha/PPM1A in Western blots. No cross-reactivity with recombinant human PP2C beta , ILKAP/PP2C delta , or PP2C gamma is observed. | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot | |
| 498922 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| EC 3.1.3.16, FLJ42306, MGC9201, MMPa-2, MPPa-1, phosphatase 2C alpha, Pp2c1, PP2CA, PP2Calpha, PP2C-ALPHA, PPM1A, PPPM1A, protein phosphatase 1A, protein phosphatase 1A (formerly 2C), magnesium-dependent, alpha isoform, protein phosphatase 2C alpha isoform, Protein phosphatase 2C isoform alpha, Protein phosphatase IA, protein phosphatase, Mg2+/Mn2+ dependent, 1A | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 5494 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction