missing translation for 'onlineSavingsMsg'
Learn More

Human/Mouse/Rat Phospho-Chk1 (S317) Antibody, R&D Systems™
GREENER_CHOICE

Product Code. 18099871
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18099871 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18099871 Supplier R&D Systems Supplier No. AF2054

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody has been used in 7 publications

Chk1 Polyclonal specifically detects Chk1 in Human, Mouse, Rat samples. It is validated for Western Blot, Simple Western.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Chk1 (p Ser317)
Applications Western Blot
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 1 ug/mL, Simple Western 10 ug/mL
Gene Alias Checkpoint, S. pombe, homolog of, 1, CHEK1, CHK1 (checkpoint, S.pombe) homolog, CHK1 checkpoint homolog (S. pombe), CHK1serine/threonine-protein kinase Chk1, EC 2.7.11, EC 2.7.11.1
Gene Symbols CHEK1
Host Species Rabbit
Immunogen Phosphopeptide containing human Chk1 S317 site
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1111
Test Specificity Detects human, mouse, and rat Chk1 when phosphorylated at S317 in Western blots. Does not recognize Chk1 when unphosphorylated at S317.
Reconstitution Reconstitute at 0.2 mg/mL in sterile PBS.
Target Species Human, Mouse, Rat
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.