missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human/Mouse/Rat p27/Kip1 Antibody, R&D Systems™

Rat Monoclonal Antibody
Brand: R&D Systems MAB22561
This item is not returnable.
View return policy
Description
p27/Kip1 Monoclonal specifically detects p27/Kip1 in Human, Mouse, Rat samples. It is validated for Immunohistochemistry.
Specifications
| p27/Kip1 | |
| Monoclonal | |
| LYOPH | |
| Immunohistochemistry 8-25 ug/mL | |
| P46527 | |
| CDKN1B | |
| E. coli-derived recombinant human p27/Kip1 Met1-Thr198 Accession # P46527 | |
| 100 μg | |
| Primary | |
| Detects human p27/Kip1 in direct ELISAs. Specificity was demonstrated by comparing staining on tissue sections of normal versus p27/Kip1 knockout mice. | |
| Human, Mouse, Rat | |
| Purified |
| Immunohistochemistry | |
| 341116 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| CDKN1B, CDKN4, cyclin-dependent kinase inhibitor 1B, cyclin-dependent kinase inhibitor 1B (p27, Kip1), Cyclin-dependent kinase inhibitor p27, Kip1, KIP1P27KIP1, MEN1B, MEN4, p27Kip1 | |
| Rat | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 1027 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction