missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human/Mouse/Rat Olig2 Antibody, R&D Systems™
Goat Polyclonal Antibody has been used in 54 publications
Brand: R&D Systems AF2418
This item is not returnable.
View return policy
Description
Olig2 Polyclonal specifically detects Olig2 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Chromatin Immunoprecipitation (ChIP), Immunocytochemistry.
Specifications
| Olig2 | |
| Polyclonal | |
| Unconjugated | |
| Q13516 | |
| OLIG2 | |
| E. coli-derived recombinant human Olig2 Met1-Lys323 Accession # Q13516 | |
| 100 μg | |
| Primary | |
| Detects human Olig2 in direct ELISAs and Western blots. In direct ELISAs, approximately 5% cross-reactivity with recombinant human (rh) Olig3 is observed. | |
| Human | |
| IgG |
| ChIP Assay, Immunocytochemistry, Immunohistochemistry | |
| LYOPH | |
| Western Blot 0.1 ug/mL, Immunohistochemistry 5-15 ug/mL, Chromatin Immunoprecipitation (ChIP) 5 ug/5 x 10^6 cells, Immunocytochemistry 5-15 ug/mL | |
| BHLHB1, BHLHE19, Class B basic helix-loop-helix protein 1, Class E basic helix-loop-helix protein 19, human protein kinase C-binding protein RACK17, OLIGO2, oligodendrocyte lineage transcription factor 2, oligodendrocyte transcription factor 2, oligodendrocyte-specific bHLH transcription factor 2, PRKCBP2, protein kinase C binding protein 2, Protein kinase C-binding protein 2, Protein kinase C-binding protein RACK17, RACK17, RACK17helix-loop-helix protein, class B, 1 | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 10215 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Korrigering av produktinnehåll
Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.
Produkttitel
Hittar du en möjlighet till förbättring?Dela en innehållskorrigering