missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human/Mouse PRDM16/MEL1 Antibody, R&D Systems™

Sheep Polyclonal Antibody has been used in 12 publications
Brand: R&D Systems AF6295
This item is not returnable.
View return policy
Description
PRDM16/MEL1 Polyclonal specifically detects PRDM16/MEL1 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry.
Specifications
| PRDM16/MEL1 | |
| Polyclonal | |
| Unconjugated | |
| A2A935.1 | |
| PRDM16 | |
| E. coli-derived recombinant mouse PRDM16 Lys537-Glu688 Accession # A2A935.1 | |
| 100 μg | |
| Primary | |
| Detects mouse and human PRDM16 in direct ELISAs and Western blots. | |
| Human, Mouse | |
| IgG |
| Western Blot, Immunohistochemistry | |
| LYOPH | |
| Western Blot 1 ug/mL, Immunohistochemistry 5-15 ug/mL | |
| MDS1/EVI1-like gene 1, MEL1, MEL1KIAA1675PFM13MGC166915, PFM13, PR domain containing 16, PR domain zinc finger protein 16, PR domain-containing protein 16, Transcription factor MEL1 | |
| Sheep | |
| Affinity Purified | |
| RUO | |
| 63976 | |
| Sterile PBS to a final concentration of 0.2 mg/mL. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction