missing translation for 'onlineSavingsMsg'
Learn More

Human/Mouse Kallikrein 7 Antibody, R&D Systems™
GREENER_CHOICE

Product Code. 18008921
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18008921 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18008921 Supplier R&D Systems Supplier No. AF2624

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Goat Polyclonal Antibody has been used in 6 publications

Kallikrein 7 Polyclonal specifically detects Kallikrein 7 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunoprecipitation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Kallikrein 7
Applications Western Blot, Immunohistochemistry, Immunoprecipitation
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 1 ug/mL, Immunohistochemistry 5-15 ug/mL, Immunoprecipitation 25 ug/mL
Gene Accession No. P49862
Gene Alias EC 3.4.21, EC 3.4.21.117, hK7, hSCCE, kallikrein 7 (chymotryptic, stratum corneum), kallikrein-related peptidase 7, KLK7, protease, serine, 6, PRSS6, SCCEkallikrein-7, Serine protease 6, signal protein, Stratum corneum chymotryptic enzyme
Gene Symbols KLK7
Host Species Goat
Immunogen Mouse myeloma cell line NS0-derived recombinant human Kallikrein 7 Glu23-His252 Accession # P49862
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5650
Test Specificity Detects human and mouse Kallikrein 7 in Western blots. Detects human Kallikrein 7 in direct ELISAs. In direct ELISAs, less than 2% cross-reactivity with recombinant human (rh) Kallikrein 1, rhKallikrein 3, rhKallikrein 4, rhKallikrein 5, rhKallikrein 8, rhKallikrein 9, rhKallikrein 11, rhKallikrein 13 and rhKallikrein 14 is observed.
Reconstitution Reconstitute at 0.2 mg/mL in sterile PBS.
Target Species Human, Mouse
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.