missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human/Mouse EBF-2 Antibody, R&D Systems™
Sheep Polyclonal Antibody has been used in 5 publications
Brand: R&D Systems AF7006
This item is not returnable.
View return policy
Description
EBF-2 Polyclonal specifically detects EBF-2 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry.
Specifications
| EBF-2 | |
| Polyclonal | |
| Unconjugated | |
| O08792 | |
| EBF2 | |
| E. coli-derived recombinant mouse EBF-2 Arg407-Ser519 Accession # O08792 | |
| 100 μg | |
| Primary | |
| Detects mouse EBF-2 in direct ELISAs. Detects mouse and human EBF-2 in Western blots. In direct ELISAs, less than 1% cross-reactivity with recombinant mouse (rm) EBF-1 and rmEBF-3 is observed. | |
| Human, Mouse | |
| IgG |
| Western Blot, Immunohistochemistry | |
| LYOPH | |
| Western Blot 1 ug/mL, Immunohistochemistry 5-15 ug/mL | |
| COE2, EBF2, EBF-2, Mmot1, O/E-3, OE-3 | |
| Sheep | |
| Affinity Purified | |
| RUO | |
| 64641 | |
| Sterile PBS to a final concentration of 0.2 mg/mL. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction