Learn More
Human/Mouse CDK4 Antibody, R&D Systems™
GREENER_CHOICE
Goat Polyclonal Antibody
Brand:  R&D Systems AF5254
Product Code. 18000872
  • 601.00€ / 100µg
Description
CDK4 Polyclonal specifically detects CDK4 in Human, Mouse samples. It is validated for Western Blot, Immunocytochemistry.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
CDK4
Polyclonal
Unconjugated
P11802
CDK4
E. coli-derived recombinant human CDK4 Met1-Glu303 Accession # P11802
100 μg
Primary
Detects human and mouse CDK4 in Western blots.
Human, Mouse
IgG
Western Blot, Immunocytochemistry
LYOPH
Western Blot 1 ug/mL, Simple Western 20 ug/mL, Immunocytochemistry 5-15 ug/mL
Cell division protein kinase 4, CMM3, cyclin-dependent kinase 4, EC 2.7.11, EC 2.7.11.22, melanoma cutaneous malignant, 3, MGC14458, PSK-J3, PSK-J3cell division kinase 4
Goat
Affinity Purified
RUO
1019
Reconstitute at 0.2 mg/mL in sterile PBS.
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.