missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human/Mouse c-Myc Antibody, R&D Systems™
Goat Polyclonal Antibody has been used in 3 publications
Brand: R&D Systems AF3696
This item is not returnable.
View return policy
Description
c-Myc Polyclonal specifically detects c-Myc in Human, Mouse samples. It is validated for Western Blot, Simple Western, Immunohistochemistry, Chromatin Immunoprecipitation (ChIP), Immunocytochemistry, Knockout Validated.
Specifications
| c-Myc | |
| Polyclonal | |
| Unconjugated | |
| P01106 | |
| MYC | |
| E. coli-derived recombinant human c-Myc Arg66-Asp201 Accession # P01106 | |
| 100 μg | |
| Primary | |
| Detects human c-Myc in direct ELISAs. Detects human and mouse c-Myc in Western blots. In direct ELISAs, less than 1% cross-reactivity with recombinant human (rh) L-Myc and rhN-Myc is observed. | |
| Human, Mouse | |
| IgG |
| ChIP Assay, Immunocytochemistry, Immunohistochemistry, KnockDown | |
| LYOPH | |
| Western Blot 0.5 ug/mL, Simple Western 20 ug/mL, Immunohistochemistry 5-15 ug/mL, Chromatin Immunoprecipitation (ChIP) 5 ug/5 x 10^6 cells, Immunocytochemistry 5-15 ug/mL, Knockout Validated | |
| avian myelocytomatosis viral oncogene homolog, BHLHE39, bHLHe39MRTL, Class E basic helix-loop-helix protein 39, cMyc, c-Myc, MYC, myc proto-oncogene protein, Myc2, MYCC, myc-related translation/localization regulatory factor, Niard, Nird, Proto-oncogene c-Myc, Transcription factor p64, v-myc avian myelocytomatosis viral oncogene homolog, v-myc myelocytomatosis viral oncogene homolog (avian) | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 4609 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction