missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human/Mouse ASAM Antibody, R&D Systems™

Sheep Polyclonal Antibody
Brand: R&D Systems AF7356
This item is not returnable.
View return policy
Description
ASAM Polyclonal specifically detects ASAM in Human, Mouse samples. It is validated for Western Blot, Flow Cytometry, CyTOF-ready.
Specifications
| ASAM | |
| Polyclonal | |
| Unconjugated | |
| Q8R373 | |
| CLMP | |
| Chinese hamster ovary cell line CHO-derived recombinant mouse ASAM Thr18-Ala234 Accession # Q8R373 | |
| 100 μg | |
| Primary | |
| Detects human and mouse recombinant ASAM in direct ELISAs. Detects human and mouse ASAM in Western blots. | |
| Human, Mouse | |
| IgG |
| Western Blot, Flow Cytometry, CyTOF | |
| LYOPH | |
| Western Blot 1 ug/mL, Flow Cytometry 2.5 ug/10^6 cells, CyTOF-ready | |
| ACAM, ACAMCoxsackie- and adenovirus receptor-like membrane protein, Adipocyte adhesion molecule, adipocyte-specific adhesion molecule, ASAMCAR-like membrane protein, Asp5, CLMP, CXADR-like membrane protein, FLJ22415 | |
| Sheep | |
| Affinity Purified | |
| RUO | |
| 79827 | |
| Sterile PBS to a final concentration of 0.2 mg/mL. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction