Learn More
Abnova™ Human MID2 Partial ORF (NP_036348, 371 a.a. - 460 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00011043-Q01.25ug
Additional Details : Weight : 0.00010kg
Description
The protein encoded by this gene is a member of the tripartite motif (TRIM) family. The TRIM motif includes three zinc-binding domains, a RING, a B-box type 1 and a B-box type 2, and a coiled-coil region. The protein localizes to microtubular structures in the cytoplasm. Alternate splicing of this gene results in two transcript variants encoding different isoforms. [provided by RefSeq]
Sequence: KLLEGLDYLTAPNPPSIREELCTASHDTITVHWISDDEFSISSYELQYTIFTGQANFISKSWCSWGLWPEIRKCKEAVSCSRLAGAPRGLSpecifications
NP_036348 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
35.64kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
KLLEGLDYLTAPNPPSIREELCTASHDTITVHWISDDEFSISSYELQYTIFTGQANFISKSWCSWGLWPEIRKCKEAVSCSRLAGAPRGL | |
RUO | |
MID2 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
11043 | |
MID2 (Human) Recombinant Protein (Q01) | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
FXY2/MID1/RNF60/TRIM1 | |
MID2 | |
Recombinant | |
wheat germ expression system |