missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human MID2 Partial ORF (NP_036348, 371 a.a. - 460 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
335.00€ - 508.00€
Specifications
Accession Number | NP_036348 |
---|---|
For Use With (Application) | Antibody Production, ELISA, Protein Array, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 11043 |
Molecular Weight (g/mol) | 35.64kDa |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16113596
|
Abnova™
H00011043-Q01.25UG |
25 ug |
508.00€
25µg |
Estimated Shipment: 10-05-2024
Log in to see stock. |
|||||
16103596
|
Abnova™
H00011043-Q01.10UG |
10 ug |
335.00€
10µg |
Estimated Shipment: 10-05-2024
Log in to see stock. |
|||||
Description
The protein encoded by this gene is a member of the tripartite motif (TRIM) family. The TRIM motif includes three zinc-binding domains, a RING, a B-box type 1 and a B-box type 2, and a coiled-coil region. The protein localizes to microtubular structures in the cytoplasm. Alternate splicing of this gene results in two transcript variants encoding different isoforms. [provided by RefSeq]
Sequence: KLLEGLDYLTAPNPPSIREELCTASHDTITVHWISDDEFSISSYELQYTIFTGQANFISKSWCSWGLWPEIRKCKEAVSCSRLAGAPRGLSpecifications
NP_036348 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
35.64kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
FXY2/MID1/RNF60/TRIM1 | |
MID2 | |
Recombinant | |
wheat germ expression system |
Antibody Production, ELISA, Protein Array, Western Blot | |
11043 | |
MID2 (Human) Recombinant Protein (Q01) | |
KLLEGLDYLTAPNPPSIREELCTASHDTITVHWISDDEFSISSYELQYTIFTGQANFISKSWCSWGLWPEIRKCKEAVSCSRLAGAPRGL | |
RUO | |
MID2 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |