missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human MFAP3 Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems MAB10798-SP
This item is not returnable.
View return policy
Description
MFAP3 Monoclonal antibody specifically detects MFAP3 in Human samples. It is validated for Immunohistochemistry
Specifications
| MFAP3 | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied either lyophilized or as a 0.2 μm filtered solution in PBS. | |
| DKFZp586F0219, microfibril-associated glycoprotein 3, microfibrillar-associated protein 3 | |
| Human embryonic kidney cell, HEK293-derived human MFAP3, Leu22-Met147, Accession # P55082 | |
| 25 μg | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20 to -70 °C under sterile conditions after reconstitution. | |
| IgG1 |
| Immunohistochemistry | |
| 1059216 | |
| Immunohistochemistry 5-25 ug/mL | |
| P55082 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 4238 | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction