missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Megalin/LRP2 Alexa Fluor 647-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems FAB9578R-100UG
This item is not returnable.
View return policy
Description
LRP2 Monoclonal specifically detects LRP2 in Human samples. It is validated for Flow Cytometry.
Specifications
| LRP2 | |
| Monoclonal | |
| Alexa Fluor 647 | |
| Supplied 0.2Mg/mL in a saline solution containing BSA and Sodium Azide. | |
| calcium sensor protein, DBS, DBSlow-density lipoprotein receptor-related protein 2, EC 1.1.2.3, EC 3.4.21.9, Glycoprotein 330, GP330, Heymann nephritis antigen homolog, low density lipoprotein receptor-related protein 2, LRP2, LRP-2, megalin | |
| Mouse myeloma cell line NS0-derived recombinant human Megalin/LRP2 Pro3510-Lys3964 Accession # P98164 | |
| 100 μg | |
| 4036 | |
| Human | |
| Purified |
| Flow Cytometry | |
| 545606 | |
| Flow Cytometry | |
| P98164 | |
| Mouse | |
| Protein A or G purified from ascites | |
| Primary | |
| Detects human Megalin/LRP2 in direct ELISAs. | |
| Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction