missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human MED18 Full-length ORF (AAH02694.1, 1 a.a. - 208 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00054797-P01.25ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16100763

  • 508.00€ / 25µg

Estimated Shipment: 04-05-2023
to see stock.
Add to basket

Description

Description

MED18 is a component of the Mediator complex, which is a coactivator for DNA-binding factors that activate transcription via RNA polymerase II (Sato et al., 2003 [PubMed 12584197]).[supplied by OMIM]

Sequence: MEAPPVTMMPVTGGTINMMEYLLQGSVLDHSLESLIHRLRGLCDNMEPETFLDHEMVFLLKGQQASPFVLRARRSMDRAGAPWHLRYLGQPEMGDKNRHALVRNCVDIATSENLTDFLMEMGFRMDHEFVAKGHLFRKGIMKIMVYKIFRILVPGNTDSTEALSLSYLVELSVVAPAGQDMVSDDMKNFAEQLKPLVHLEKIDPKRLM
Specifications

Specifications

AAH02694.1
Liquid
54797
MED18 (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MEAPPVTMMPVTGGTINMMEYLLQGSVLDHSLESLIHRLRGLCDNMEPETFLDHEMVFLLKGQQASPFVLRARRSMDRAGAPWHLRYLGQPEMGDKNRHALVRNCVDIATSENLTDFLMEMGFRMDHEFVAKGHLFRKGIMKIMVYKIFRILVPGNTDSTEALSLSYLVELSVVAPAGQDMVSDDMKNFAEQLKPLVHLEKIDPKRLM
RUO
MED18
Wheat Germ (in vitro)
GST
Antibody Production, Array, Enzyme-linked Immunoabsorbent Assay, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
50.1kDa
Glutathione Sepharose 4 Fast Flow
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
FLJ20045/p28b
MED18
Yes
wheat germ expression system
Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers