Learn More
Abnova™ Human MCMDC1 Partial ORF (AAH31658, 1 a.a. - 100 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00254394-Q01.25ug
Additional Details : Weight : 0.00010kg
Description
This gene encodes a protein that shares similarity with minichromosome maintenance (MCM) proteins, which are known to be essential for initiation of DNA replication. [provided by RefSeq]
Sequence: MNSDQVTLVGQVFESYVSEYHKNDILLILKERDEDAHYPVVVNAMTLFETNMEIGEYFNMFPSEVLTIFDSALRRSALTILQSLSQPEAVSMKQNLHARISpecifications
AAH31658 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.41kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MNSDQVTLVGQVFESYVSEYHKNDILLILKERDEDAHYPVVVNAMTLFETNMEIGEYFNMFPSEVLTIFDSALRRSALTILQSLSQPEAVSMKQNLHARI | |
RUO | |
MCM9 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
254394 | |
MCMDC1 (Human) Recombinant Protein (Q01) | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
C6orf61/FLJ13942/FLJ20170/FLJ56845/MCMDC1/MGC35304/dJ329L24.1/dJ329L24.3 | |
MCM9 | |
Recombinant | |
wheat germ expression system |