missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human MCMDC1 Partial ORF (AAH31658, 1 a.a. - 100 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
335.00€ - 508.00€
Specifications
Accession Number | AAH31658 |
---|---|
For Use With (Application) | Antibody Production, ELISA, Protein Array, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 254394 |
Molecular Weight (g/mol) | 36.41kDa |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16112607
|
Abnova™
H00254394-Q01.25UG |
25 ug |
508.00€
25µg |
Estimated Shipment: 05-06-2024
Log in to see stock. |
|||||
16102607
|
Abnova™
H00254394-Q01.10UG |
10 ug |
335.00€
10µg |
Estimated Shipment: 05-06-2024
Log in to see stock. |
|||||
Description
This gene encodes a protein that shares similarity with minichromosome maintenance (MCM) proteins, which are known to be essential for initiation of DNA replication. [provided by RefSeq]
Sequence: MNSDQVTLVGQVFESYVSEYHKNDILLILKERDEDAHYPVVVNAMTLFETNMEIGEYFNMFPSEVLTIFDSALRRSALTILQSLSQPEAVSMKQNLHARISpecifications
AAH31658 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.41kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
C6orf61/FLJ13942/FLJ20170/FLJ56845/MCMDC1/MGC35304/dJ329L24.1/dJ329L24.3 | |
MCM9 | |
Recombinant | |
wheat germ expression system |
Antibody Production, ELISA, Protein Array, Western Blot | |
254394 | |
MCMDC1 (Human) Recombinant Protein (Q01) | |
MNSDQVTLVGQVFESYVSEYHKNDILLILKERDEDAHYPVVVNAMTLFETNMEIGEYFNMFPSEVLTIFDSALRRSALTILQSLSQPEAVSMKQNLHARI | |
RUO | |
MCM9 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |