missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human MAGEA5 Partial ORF (NP_066387.1, 60 a.a. - 124 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00004104-Q01.10ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16120945

  • 359.00€ / 10µg

Estimated Shipment: 27-09-2022
to see stock.
Add to basket

Description

Description

This gene is a member of the MAGEA gene family. The members of this family encode proteins with 50 to 80% sequence identity to each other. The promoters and first exons of the MAGEA genes show considerable variability, suggesting that the existence of this gene family enables the same function to be expressed under different transcriptional controls. The MAGEA genes are clustered at chromosomal location Xq28. They have been implicated in some hereditary disorders, such as dyskeratosis congenita. [provided by RefSeq]

Sequence: GPLKSPQGASAIPTAIDFTLWRQSIKGSSNQEEEGPSTSPDPESVFRAALSKKVADLIHFLLLKY
Specifications

Specifications

NP_066387.1
Liquid
4104
MAGEA5 (Human) Recombinant Protein (Q01)
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
MAGE5/MAGEA4/MGC129526
MAGEA5
Yes
wheat germ expression system
Antibody Production, Enzyme-linked Immunoabsorbent Assay, Protein Array, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
32.89kDa
12.5% SDS-PAGE Stained with Coomassie Blue
GPLKSPQGASAIPTAIDFTLWRQSIKGSSNQEEEGPSTSPDPESVFRAALSKKVADLIHFLLLKY
RUO
MAGEA5
Wheat Germ (in vitro)
GST
Videos
SDS
Documents
Certificates
Special Offers

Special Offers