missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human MAGEA5 Partial ORF (NP_066387.1, 60 a.a. - 124 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
335.00€ - 508.00€
Specifications
Accession Number | NP_066387.1 |
---|---|
For Use With (Application) | Antibody Production, ELISA, Protein Array, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 4104 |
Molecular Weight (g/mol) | 32.89kDa |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16130945
|
Abnova™
H00004104-Q01.25UG |
25 ug |
508.00€
25µg |
Estimated Shipment: 31-05-2024
Log in to see stock. |
|||||
16120945
|
Abnova™
H00004104-Q01.10UG |
10 ug |
335.00€
10µg |
Estimated Shipment: 31-05-2024
Log in to see stock. |
|||||
Description
This gene is a member of the MAGEA gene family. The members of this family encode proteins with 50 to 80% sequence identity to each other. The promoters and first exons of the MAGEA genes show considerable variability, suggesting that the existence of this gene family enables the same function to be expressed under different transcriptional controls. The MAGEA genes are clustered at chromosomal location Xq28. They have been implicated in some hereditary disorders, such as dyskeratosis congenita. [provided by RefSeq]
Sequence: GPLKSPQGASAIPTAIDFTLWRQSIKGSSNQEEEGPSTSPDPESVFRAALSKKVADLIHFLLLKYSpecifications
NP_066387.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
32.89kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
MAGE5/MAGEA4/MGC129526 | |
MAGEA5 | |
Recombinant | |
wheat germ expression system |
Antibody Production, ELISA, Protein Array, Western Blot | |
4104 | |
MAGEA5 (Human) Recombinant Protein (Q01) | |
GPLKSPQGASAIPTAIDFTLWRQSIKGSSNQEEEGPSTSPDPESVFRAALSKKVADLIHFLLLKY | |
RUO | |
MAGEA5 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |