Learn More
Abnova™ Human LLGL1 Partial ORF (NP_004131, 911 a.a. - 1010 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00003996-Q01.25ug
Additional Details : Weight : 0.00010kg
Description
This gene encodes a protein that is similar to a tumor suppressor in Drosophila. The protein is part of a cytoskeletal network and is associated with nonmuscle myosin II heavy chain and a kinase that specifically phosphorylates this protein at serine residues. The gene is located within the Smith-Magenis syndrome region on chromosome 17. [provided by RefSeq]
Sequence: GIASCVFTRHGQGFYLISPSEFERFSLSARNITEPLCSLDINWPRDATQASYRIRESPKLSQANGTPSILLAPQSLDGSPDPAHSMGPDTPEPPEAALSPSpecifications
NP_004131 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.74kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
GIASCVFTRHGQGFYLISPSEFERFSLSARNITEPLCSLDINWPRDATQASYRIRESPKLSQANGTPSILLAPQSLDGSPDPAHSMGPDTPEPPEAALSP | |
RUO | |
LLGL1 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
3996 | |
LLGL1 (Human) Recombinant Protein (Q01) | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
DLG4/HUGL/HUGL-1/HUGL1/LLGL | |
LLGL1 | |
Recombinant | |
wheat germ expression system |