missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human LLGL1 Partial ORF (NP_004131, 911 a.a. - 1010 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
335.00€ - 508.00€
Specifications
Accession Number | NP_004131 |
---|---|
For Use With (Application) | Antibody Production, ELISA, Protein Array, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 3996 |
Molecular Weight (g/mol) | 36.74kDa |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16170725
|
Abnova™
H00003996-Q01.25UG |
25 ug |
508.00€
25µg |
Estimated Shipment: 31-05-2024
Log in to see stock. |
|||||
16160725
|
Abnova™
H00003996-Q01.10UG |
10 ug |
335.00€
10µg |
Estimated Shipment: 31-05-2024
Log in to see stock. |
|||||
Description
This gene encodes a protein that is similar to a tumor suppressor in Drosophila. The protein is part of a cytoskeletal network and is associated with nonmuscle myosin II heavy chain and a kinase that specifically phosphorylates this protein at serine residues. The gene is located within the Smith-Magenis syndrome region on chromosome 17. [provided by RefSeq]
Sequence: GIASCVFTRHGQGFYLISPSEFERFSLSARNITEPLCSLDINWPRDATQASYRIRESPKLSQANGTPSILLAPQSLDGSPDPAHSMGPDTPEPPEAALSPSpecifications
NP_004131 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.74kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
DLG4/HUGL/HUGL-1/HUGL1/LLGL | |
LLGL1 | |
Recombinant | |
wheat germ expression system |
Antibody Production, ELISA, Protein Array, Western Blot | |
3996 | |
LLGL1 (Human) Recombinant Protein (Q01) | |
GIASCVFTRHGQGFYLISPSEFERFSLSARNITEPLCSLDINWPRDATQASYRIRESPKLSQANGTPSILLAPQSLDGSPDPAHSMGPDTPEPPEAALSP | |
RUO | |
LLGL1 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |