Learn More
Abnova™ Human LHFPL4 Full-length ORF (NP_940962.1, 1 a.a. - 247 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00375323-P01.10ug
Additional Details : Weight : 0.00010kg
Description
This gene is a member of the lipoma HMGIC fusion partner (LHFP) gene family, which is a subset of the superfamily of tetraspan transmembrane protein encoding genes. Mutations in one LHFP-like gene result in deafness in humans and mice, and a second LHFP-like gene is fused to a high-mobility group gene in a translocation-associated lipoma. [provided by RefSeq]
Sequence: MLPSQEASKLYHEHYMRNSRAIGVLWAIFTICFAIINVVVFIQPYWVGDSVSTPKPGYFGLFHYCVGSGLAGRELTCRGSFTDFSTIPSSAFKAAAFFVLLSMVLILGCITCFSLFFFCNTATVYKICAWMQLLAALCLVLGCMIFPDGWDAETIRDMCGAKTGKYSLGDCSVRWAYILAIIGILNALILSFLAFVLGNRQTDLLQEELKPENKDFVGSTVSSVLRPGGDVSGWGVLPCPVAHSQGPSpecifications
NP_940962.1 | |
Liquid | |
375323 | |
LHFPL4 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MLPSQEASKLYHEHYMRNSRAIGVLWAIFTICFAIINVVVFIQPYWVGDSVSTPKPGYFGLFHYCVGSGLAGRELTCRGSFTDFSTIPSSAFKAAAFFVLLSMVLILGCITCFSLFFFCNTATVYKICAWMQLLAALCLVLGCMIFPDGWDAETIRDMCGAKTGKYSLGDCSVRWAYILAIIGILNALILSFLAFVLGNRQTDLLQEELKPENKDFVGSTVSSVLRPGGDVSGWGVLPCPVAHSQGP | |
RUO | |
LHFPL4 | |
Wheat Germ (in vitro) | |
GST |
Antibody Production, Array, Enzyme-linked Immunoabsorbent Assay, Western Blot (Recombinant protein) | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
53.4kDa | |
Glutathione Sepharose 4 Fast Flow | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
MGC133162 | |
LHFPL4 | |
Yes | |
wheat germ expression system |