missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human LHFPL4 Full-length ORF (NP_940962.1, 1 a.a. - 247 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
335.00€ - 508.00€
Specifications
Accession Number | NP_940962.1 |
---|---|
For Use With (Application) | Antibody Production, Protein Array, ELISA, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 375323 |
Molecular Weight (g/mol) | 53.4kDa |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16170394
|
Abnova™
H00375323-P01.25UG |
25 ug |
508.00€
25µg |
Estimated Shipment: 31-05-2024
Log in to see stock. |
|||||
16160394
|
Abnova™
H00375323-P01.10UG |
10 ug |
335.00€
10µg |
Estimated Shipment: 31-05-2024
Log in to see stock. |
|||||
Description
This gene is a member of the lipoma HMGIC fusion partner (LHFP) gene family, which is a subset of the superfamily of tetraspan transmembrane protein encoding genes. Mutations in one LHFP-like gene result in deafness in humans and mice, and a second LHFP-like gene is fused to a high-mobility group gene in a translocation-associated lipoma. [provided by RefSeq]
Sequence: MLPSQEASKLYHEHYMRNSRAIGVLWAIFTICFAIINVVVFIQPYWVGDSVSTPKPGYFGLFHYCVGSGLAGRELTCRGSFTDFSTIPSSAFKAAAFFVLLSMVLILGCITCFSLFFFCNTATVYKICAWMQLLAALCLVLGCMIFPDGWDAETIRDMCGAKTGKYSLGDCSVRWAYILAIIGILNALILSFLAFVLGNRQTDLLQEELKPENKDFVGSTVSSVLRPGGDVSGWGVLPCPVAHSQGPSpecifications
NP_940962.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
53.4kDa | |
Glutathione Sepharose 4 Fast Flow | |
MLPSQEASKLYHEHYMRNSRAIGVLWAIFTICFAIINVVVFIQPYWVGDSVSTPKPGYFGLFHYCVGSGLAGRELTCRGSFTDFSTIPSSAFKAAAFFVLLSMVLILGCITCFSLFFFCNTATVYKICAWMQLLAALCLVLGCMIFPDGWDAETIRDMCGAKTGKYSLGDCSVRWAYILAIIGILNALILSFLAFVLGNRQTDLLQEELKPENKDFVGSTVSSVLRPGGDVSGWGVLPCPVAHSQGP | |
RUO | |
LHFPL4 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, Protein Array, ELISA, Western Blot | |
375323 | |
LHFPL4 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
MGC133162 | |
LHFPL4 | |
Recombinant | |
wheat germ expression system |