missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human LHFPL4 Full-length ORF (NP_940962.1, 1 a.a. - 247 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00375323-P01.25ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16170394

  • 508.00€ / 25µg
Estimated Shipment: 12-06-2024
to see stock.
Add to basket

Description

Description

This gene is a member of the lipoma HMGIC fusion partner (LHFP) gene family, which is a subset of the superfamily of tetraspan transmembrane protein encoding genes. Mutations in one LHFP-like gene result in deafness in humans and mice, and a second LHFP-like gene is fused to a high-mobility group gene in a translocation-associated lipoma. [provided by RefSeq]

Sequence: MLPSQEASKLYHEHYMRNSRAIGVLWAIFTICFAIINVVVFIQPYWVGDSVSTPKPGYFGLFHYCVGSGLAGRELTCRGSFTDFSTIPSSAFKAAAFFVLLSMVLILGCITCFSLFFFCNTATVYKICAWMQLLAALCLVLGCMIFPDGWDAETIRDMCGAKTGKYSLGDCSVRWAYILAIIGILNALILSFLAFVLGNRQTDLLQEELKPENKDFVGSTVSSVLRPGGDVSGWGVLPCPVAHSQGP
Specifications

Specifications

NP_940962.1
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
53.4kDa
Glutathione Sepharose 4 Fast Flow
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
MGC133162
LHFPL4
Recombinant
wheat germ expression system
Antibody Production, Protein Array, ELISA, Western Blot
375323
LHFPL4 (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MLPSQEASKLYHEHYMRNSRAIGVLWAIFTICFAIINVVVFIQPYWVGDSVSTPKPGYFGLFHYCVGSGLAGRELTCRGSFTDFSTIPSSAFKAAAFFVLLSMVLILGCITCFSLFFFCNTATVYKICAWMQLLAALCLVLGCMIFPDGWDAETIRDMCGAKTGKYSLGDCSVRWAYILAIIGILNALILSFLAFVLGNRQTDLLQEELKPENKDFVGSTVSSVLRPGGDVSGWGVLPCPVAHSQGP
RUO
LHFPL4
Wheat Germ (in vitro)
GST
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers