missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human LARS2 Partial ORF (NP_056155, 806 a.a. - 903 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00023395-Q01.10ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16107252

  • 349.00€ / 10µg

Estimated Shipment: 17-07-2023
to see stock.
Add to basket

Description

Description

This gene encodes a class 1 aminoacyl-tRNA synthetase, mitochondrial leucyl-tRNA synthetase. Each of the twenty aminoacyl-tRNA synthetases catalyzes the aminoacylation of a specific tRNA or tRNA isoaccepting family with the cognate amino acid. [provided by RefSeq]

Sequence: VPRKLCAHYTWDASVLLQAWPAVDPEFLQQPEVVQMAVLINNKACGKIPVPQQVARDQDKVHEFVLQSELGVRLLQGRSIKKSFLSPRTALINFLVQD
Specifications

Specifications

NP_056155
Liquid
23395
LARS2 (Human) Recombinant Protein (Q01)
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
KIAA0028/LEURS/MGC26121
LARS2
Yes
wheat germ expression system
Antibody Production, Enzyme-linked Immunoabsorbent Assay, Protein Array, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
36.52kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
VPRKLCAHYTWDASVLLQAWPAVDPEFLQQPEVVQMAVLINNKACGKIPVPQQVARDQDKVHEFVLQSELGVRLLQGRSIKKSFLSPRTALINFLVQD
RUO
LARS2
Wheat Germ (in vitro)
GST
Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers