missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human LARP2 Full-length ORF (AAH30516.1, 1 a.a. - 201 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00055132-P01.25ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16121153

  • 508.00€ / 25µg
Estimated Shipment: 11-06-2024
to see stock.
Add to basket

Description

Description

This gene encodes a protein containing domains found in the La related protein of Drosophila melanogaster. La motif-containing proteins are thought to be RNA-binding proteins, where the La motif and adjacent amino acids fold into an RNA recognition motif. The La motif is also found in proteins unrelated to the La protein. Alternative splicing has been observed at this locus and three variants, encoding distinct isoforms, are described. Additional splice variation has been identified but the full-length nature of these transcripts has not been determined. [provided by RefSeq]

Sequence: MDSRDHGPGTSSVSTSNASPSEGAPLAGSYGCTPHSFPKFQHPSHELLKENGFTQQVYHKYRRRCLSERKRLGIGQSQEMNTLFRFWSFFLRDHFNKKMYEEFRQLAWEDAKENYRYGLECLFRFYSYGLEKKFRREIFQDFQEETKKDYESGQLYGLEKFWAYLKYSQSKTQSIDPKLQEYLCSFKRLEDFRVDEADPIE
Specifications

Specifications

AAH30516.1
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
50.5kDa
Glutathione Sepharose 4 Fast Flow
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
DKFZp686L13217/MGC117277/MGC75174
LARP2
Recombinant
wheat germ expression system
Antibody Production, Protein Array, ELISA, Western Blot
55132
LARP2 (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MDSRDHGPGTSSVSTSNASPSEGAPLAGSYGCTPHSFPKFQHPSHELLKENGFTQQVYHKYRRRCLSERKRLGIGQSQEMNTLFRFWSFFLRDHFNKKMYEEFRQLAWEDAKENYRYGLECLFRFYSYGLEKKFRREIFQDFQEETKKDYESGQLYGLEKFWAYLKYSQSKTQSIDPKLQEYLCSFKRLEDFRVDEADPIE
RUO
LARP2
Wheat Germ (in vitro)
GST
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers