Learn More
Human LAP (TGF-beta 1) Antibody, R&D Systems™
Goat Polyclonal Antibody has been used in 17 publications
Brand: R&D Systems AF-246-NA
This item is not returnable.
View return policy
Description
LAP (TGF-beta 1) Polyclonal specifically detects LAP (TGF-beta 1) in Human samples. It is validated for Western Blot, Simple Western, Neutralization, Immunocytochemistry.
Specifications
| LAP (TGF-beta 1) | |
| Polyclonal | |
| Unconjugated | |
| P01137 | |
| TGFB1 | |
| S. frugiperda insect ovarian cell line Sf 21-derived and Chinese hamster ovary cell line CHO-derived recombinant human LAP TGF-beta 1 Leu30-Ser390 Accession # P01137 | |
| 100 μg | |
| Primary | |
| Detects human LAP TGF-beta 1 in direct ELISAs and Western blots. In direct ELISAs and Western blots, this antibody is specific for the LAP derived from the TGF-beta 1 precursor, but does not react with LAP from the TGF-beta 2 precursor. | |
| Human | |
| IgG |
| Western Blot, Neutralization, Immunocytochemistry | |
| LYOPH | |
| Western Blot 2 ug/mL, Simple Western 20 ug/mL, Neutralization 0.4-2 ug/mL, Immunocytochemistry 5-15 ug/mL | |
| CED, DPD1, LAP, LAP (TGFbeta 1), TGFB, TGFB1, TGFbeta, transforming growth factor beta 1 | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 7040 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |