missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human KRT79 Full-length ORF (NP_787028.1, 1 a.a. - 535 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00338785-P01.10ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16120134

  • 335.00€ / 10µg
Estimated Shipment: 11-06-2024
to see stock.
Add to basket

Description

Description

Keratins are intermediate filament proteins responsible for the structural integrity of epithelial cells and are subdivided into epithelial keratins and hair keratins. This gene encodes an epithelial keratin that is expressed in skeletal muscle, skin and scalp. The type II keratins are clustered in a region of chromosome 12q13

Sequence: MRSSVSRQTYSTKGGFSSNSASGGSGSQARTSFSSVTVSRSSGSGGGAHCGPGTGGFGSRSLYNLGGHKSISVSVAGGALLGRALGGFGFGSRAFMGQGAGRQTFGPACPPGGIQEVTVNQSLLTPLHVEIDPEIQRVRTQEREQIKTLNNKFASFIDKVRFLEQQNKVLETKWALLQEQGQNLGVTRNNLEPLFEAYLGSMRSTLDRLQSERGRLDSELRNVQDLVEDFKNKYEDEINKHTAAENEFVVLKKDVDAAYMGRMDLHGKVGTLTQEIDFLQQLYEMELSQVQTHVSNTNVVLSMDNNRNLDLDSIIAEVKAQYELIAQRSRAEAEAWYQTKYEELQVTAGKHGDNLRDTKNEIAELTRTIQRLQGEADAAKKQCQQLQTAIAEAEQRGELALKDAQKKLGDLDVALHQAKEDLTRLLRDYQELMNVKLALDVEIATYRKLLESEESRMSGECPSAVSISVTGNSTTVCGGGAASFGGGISLGGSGGATKGGFSTNVGYSTVKGGPVSAGTSILRKTTTVKTSSQRY
Specifications

Specifications

NP_787028.1
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
84.2kDa
Glutathione Sepharose 4 Fast Flow
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
FLJ26646/K6L/KRT6L
KRT79
Recombinant
wheat germ expression system
Antibody Production, Protein Array, ELISA, Western Blot
338785
KRT79 (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MRSSVSRQTYSTKGGFSSNSASGGSGSQARTSFSSVTVSRSSGSGGGAHCGPGTGGFGSRSLYNLGGHKSISVSVAGGALLGRALGGFGFGSRAFMGQGAGRQTFGPACPPGGIQEVTVNQSLLTPLHVEIDPEIQRVRTQEREQIKTLNNKFASFIDKVRFLEQQNKVLETKWALLQEQGQNLGVTRNNLEPLFEAYLGSMRSTLDRLQSERGRLDSELRNVQDLVEDFKNKYEDEINKHTAAENEFVVLKKDVDAAYMGRMDLHGKVGTLTQEIDFLQQLYEMELSQVQTHVSNTNVVLSMDNNRNLDLDSIIAEVKAQYELIAQRSRAEAEAWYQTKYEELQVTAGKHGDNLRDTKNEIAELTRTIQRLQGEADAAKKQCQQLQTAIAEAEQRGELALKDAQKKLGDLDVALHQAKEDLTRLLRDYQELMNVKLALDVEIATYRKLLESEESRMSGECPSAVSISVTGNSTTVCGGGAASFGGGISLGGSGGATKGGFSTNVGYSTVKGGPVSAGTSILRKTTTVKTSSQRY
RUO
KRT79
Wheat Germ (in vitro)
GST
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers