missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human KRT79 Full-length ORF (NP_787028.1, 1 a.a. - 535 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
335.00€ - 508.00€
Specifications
Accession Number | NP_787028.1 |
---|---|
For Use With (Application) | Antibody Production, Protein Array, ELISA, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 338785 |
Molecular Weight (g/mol) | 84.2kDa |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16130134
|
Abnova™
H00338785-P01.25UG |
25 ug |
508.00€
25µg |
Estimated Shipment: 31-05-2024
Log in to see stock. |
|||||
16120134
|
Abnova™
H00338785-P01.10UG |
10 ug |
335.00€
10µg |
Estimated Shipment: 31-05-2024
Log in to see stock. |
|||||
Description
Keratins are intermediate filament proteins responsible for the structural integrity of epithelial cells and are subdivided into epithelial keratins and hair keratins. This gene encodes an epithelial keratin that is expressed in skeletal muscle, skin and scalp. The type II keratins are clustered in a region of chromosome 12q13
Sequence: MRSSVSRQTYSTKGGFSSNSASGGSGSQARTSFSSVTVSRSSGSGGGAHCGPGTGGFGSRSLYNLGGHKSISVSVAGGALLGRALGGFGFGSRAFMGQGAGRQTFGPACPPGGIQEVTVNQSLLTPLHVEIDPEIQRVRTQEREQIKTLNNKFASFIDKVRFLEQQNKVLETKWALLQEQGQNLGVTRNNLEPLFEAYLGSMRSTLDRLQSERGRLDSELRNVQDLVEDFKNKYEDEINKHTAAENEFVVLKKDVDAAYMGRMDLHGKVGTLTQEIDFLQQLYEMELSQVQTHVSNTNVVLSMDNNRNLDLDSIIAEVKAQYELIAQRSRAEAEAWYQTKYEELQVTAGKHGDNLRDTKNEIAELTRTIQRLQGEADAAKKQCQQLQTAIAEAEQRGELALKDAQKKLGDLDVALHQAKEDLTRLLRDYQELMNVKLALDVEIATYRKLLESEESRMSGECPSAVSISVTGNSTTVCGGGAASFGGGISLGGSGGATKGGFSTNVGYSTVKGGPVSAGTSILRKTTTVKTSSQRYSpecifications
NP_787028.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
84.2kDa | |
Glutathione Sepharose 4 Fast Flow | |
MRSSVSRQTYSTKGGFSSNSASGGSGSQARTSFSSVTVSRSSGSGGGAHCGPGTGGFGSRSLYNLGGHKSISVSVAGGALLGRALGGFGFGSRAFMGQGAGRQTFGPACPPGGIQEVTVNQSLLTPLHVEIDPEIQRVRTQEREQIKTLNNKFASFIDKVRFLEQQNKVLETKWALLQEQGQNLGVTRNNLEPLFEAYLGSMRSTLDRLQSERGRLDSELRNVQDLVEDFKNKYEDEINKHTAAENEFVVLKKDVDAAYMGRMDLHGKVGTLTQEIDFLQQLYEMELSQVQTHVSNTNVVLSMDNNRNLDLDSIIAEVKAQYELIAQRSRAEAEAWYQTKYEELQVTAGKHGDNLRDTKNEIAELTRTIQRLQGEADAAKKQCQQLQTAIAEAEQRGELALKDAQKKLGDLDVALHQAKEDLTRLLRDYQELMNVKLALDVEIATYRKLLESEESRMSGECPSAVSISVTGNSTTVCGGGAASFGGGISLGGSGGATKGGFSTNVGYSTVKGGPVSAGTSILRKTTTVKTSSQRY | |
RUO | |
KRT79 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, Protein Array, ELISA, Western Blot | |
338785 | |
KRT79 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
FLJ26646/K6L/KRT6L | |
KRT79 | |
Recombinant | |
wheat germ expression system |