missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Kininogen Light Chain Antibody, R&D Systems™

Mouse Monoclonal Antibody
196.00€ - 530.00€
Specifications
| Antigen | Kininogen |
|---|---|
| Clone | 236006 |
| Concentration | LYOPH |
| Dilution | Western Blot 1 ug/mL, Immunoprecipitation 25 ug/mL |
| Applications | Western Blot, Immunoprecipitation |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18783909
![]() |
R&D Systems
MAB15691-SP |
25 μg |
196.00€
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18190601
![]() |
R&D Systems
MAB15691 |
500 μg |
530.00€
500µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Kininogen Monoclonal specifically detects Kininogen in Human samples. It is validated for Western Blot, Immunoprecipitation.Specifications
| Kininogen | |
| LYOPH | |
| Western Blot, Immunoprecipitation | |
| Unconjugated | |
| Mouse | |
| Human | |
| P01042.2 | |
| KNG1 | |
| IgG2b | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| 236006 | |
| Western Blot 1 ug/mL, Immunoprecipitation 25 ug/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| 3827 | |
| Mouse myeloma cell line NS0-derived recombinant human Kininogen Gln19-Ser644 (Ile581Thr) (predicted) Accession # P01042.2 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Detects human Kininogen Light Chain in direct ELISAs and Western blots. In Western blots, it reacts with the full length (amino acids 19-644) and two forms of the light chain starting at amino acids 390 and 438, respectively, and does not react with the E. coli-expressed D5/Kininostatin. In Western blots, no cross-reactivity with recombinant human (rh) Cystatins A, B, C, D, E/M, F, S, SA, SN, rhFetuins A, B, rhHPRG, or recombinant mouse Kininogen is observed. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title