Learn More
Abnova™ Human KIFAP3 Full-length ORF (ABM85040.1, 1 a.a. - 792 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
335.00€ - 508.00€
Specifications
Accession Number | ABM85040.1 |
---|---|
For Use With (Application) | Antibody Production, Protein Array, ELISA, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 22920 |
Molecular Weight (g/mol) | 113.52kDa |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16166742
|
Abnova™
H00022920-P01.25UG |
25 ug |
508.00€
25µg |
Estimated Shipment: 06-06-2024
Log in to see stock. |
|||||
16156742
|
Abnova™
H00022920-P01.10UG |
10 ug |
335.00€
10µg |
Estimated Shipment: 06-06-2024
Log in to see stock. |
|||||
Description
The small G protein GDP dissociation stimulator (smg GDS) is a regulator protein having two activities on a group of small G proteins including the Rho and Rap1 family members and Ki-Ras; one is to stimulate their GDP/GTP exchange reactions, and the other is to inhibit their interactions with membranes. The protein encoded by this gene contains 9 'Armadillo' repeats and interacts with the smg GDS protein through these repeats. This protein, which is highly concentrated around the endoplasmic reticulum, is phosphorylated by v-src, and this phosphorylation reduces the affinity of the protein for smg GDS. It is thought that this protein serves as a linker between human chromosome-associated polypeptide (HCAP) and KIF3A/B, a kinesin superfamily protein in the nucleus, and that it plays a role in the interaction of chromosomes with an ATPase motor protein. [provided by RefSeq]
Sequence: MQGEDARYLKRKVKGGNIDVHPSEKALIVHYEVEATILGEMGDPMLGERKECQKIIRLKSLNANTDITSLARKVVEECKLIHPSKLNEVEQLLYYLQNRRDSLSGKEKKEKSSKPKDPPPFEGMEIDEVANINDMDEYIELLYEDIPDKVRGSALILQLARNPDNLEELLLNETALGALARVLREDWKQSVELATNIIYIFFCFSSFSQFHGLITHYKIGALCMNIIDHELKRHELWQEELSKKKKAVDEDPENQTLRKDYEKTFKKYQGLVVKQEQLLRVALYLLLNLAEDTRTELKMRNKNIVHMLVKALDRDNFELLILVVSFLKKLSIFMENKNDMVEMDIVEKLVKMIPCEHEDLLNITLRLLLNLSFDTGLRNKMVQVGLLPKLTALLGNDNYKQIAMCVLYHISMDDRFKSMFAYTDCIPQLMKMLFECSDERIDLELISFCINLAANKRNVQLICEGNGLKMLMKRALKFKDPLLMKMIRNISQHDGPTKNLFIDYVGDLAAQISNDEEEEFVIECLGTLANLTIPDLDWELVLKEYKLVPYLKDKLKPGAAEDDLVLEVVIMIGTVSMDDSCAALLAKSGIIPALIELLNAQQEDDEFVCQIIYVFYQMVFHQATRDVIIKETQAPAYLIDLMHDKNNEIRKVCDNTLDIIAEYDEEWAKKIQSEKFRWHNSQWLEMVESRQMDESEQYLYGDDRIEPYIHEGDILERPDLFYNSDGLIASEGAISPDFFNDYHLQNGDVVGQHSFPGSLGMDGFGQPVGILGRPATAYGFRPDEPYYYGYGSSpecifications
ABM85040.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
113.52kDa | |
Glutathione Sepharose 4 Fast Flow | |
MQGEDARYLKRKVKGGNIDVHPSEKALIVHYEVEATILGEMGDPMLGERKECQKIIRLKSLNANTDITSLARKVVEECKLIHPSKLNEVEQLLYYLQNRRDSLSGKEKKEKSSKPKDPPPFEGMEIDEVANINDMDEYIELLYEDIPDKVRGSALILQLARNPDNLEELLLNETALGALARVLREDWKQSVELATNIIYIFFCFSSFSQFHGLITHYKIGALCMNIIDHELKRHELWQEELSKKKKAVDEDPENQTLRKDYEKTFKKYQGLVVKQEQLLRVALYLLLNLAEDTRTELKMRNKNIVHMLVKALDRDNFELLILVVSFLKKLSIFMENKNDMVEMDIVEKLVKMIPCEHEDLLNITLRLLLNLSFDTGLRNKMVQVGLLPKLTALLGNDNYKQIAMCVLYHISMDDRFKSMFAYTDCIPQLMKMLFECSDERIDLELISFCINLAANKRNVQLICEGNGLKMLMKRALKFKDPLLMKMIRNISQHDGPTKNLFIDYVGDLAAQISNDEEEEFVIECLGTLANLTIPDLDWELVLKEYKLVPYLKDKLKPGAAEDDLVLEVVIMIGTVSMDDSCAALLAKSGIIPALIELLNAQQEDDEFVCQIIYVFYQMVFHQATRDVIIKETQAPAYLIDLMHDKNNEIRKVCDNTLDIIAEYDEEWAKKIQSEKFRWHNSQWLEMVESRQMDESEQYLYGDDRIEPYIHEGDILERPDLFYNSDGLIASEGAISPDFFNDYHLQNGDVVGQHSFPGSLGMDGFGQPVGILGRPATAYGFRPDEPYYYGYGS | |
RUO | |
KIFAP3 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, Protein Array, ELISA, Western Blot | |
22920 | |
KIFAP3 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
FLJ22818/KAP3/SMAP/Smg-GDS/dJ190I16.1 | |
KIFAP3 | |
Recombinant | |
wheat germ expression system |