missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human KIFAP3 Full-length ORF (ABM85040.1, 1 a.a. - 792 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00022920-P01.10ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16156742

  • 335.00€ / 10µg
Estimated Shipment: 14-06-2024
to see stock.
Add to basket

Description

Description

The small G protein GDP dissociation stimulator (smg GDS) is a regulator protein having two activities on a group of small G proteins including the Rho and Rap1 family members and Ki-Ras; one is to stimulate their GDP/GTP exchange reactions, and the other is to inhibit their interactions with membranes. The protein encoded by this gene contains 9 'Armadillo' repeats and interacts with the smg GDS protein through these repeats. This protein, which is highly concentrated around the endoplasmic reticulum, is phosphorylated by v-src, and this phosphorylation reduces the affinity of the protein for smg GDS. It is thought that this protein serves as a linker between human chromosome-associated polypeptide (HCAP) and KIF3A/B, a kinesin superfamily protein in the nucleus, and that it plays a role in the interaction of chromosomes with an ATPase motor protein. [provided by RefSeq]

Sequence: MQGEDARYLKRKVKGGNIDVHPSEKALIVHYEVEATILGEMGDPMLGERKECQKIIRLKSLNANTDITSLARKVVEECKLIHPSKLNEVEQLLYYLQNRRDSLSGKEKKEKSSKPKDPPPFEGMEIDEVANINDMDEYIELLYEDIPDKVRGSALILQLARNPDNLEELLLNETALGALARVLREDWKQSVELATNIIYIFFCFSSFSQFHGLITHYKIGALCMNIIDHELKRHELWQEELSKKKKAVDEDPENQTLRKDYEKTFKKYQGLVVKQEQLLRVALYLLLNLAEDTRTELKMRNKNIVHMLVKALDRDNFELLILVVSFLKKLSIFMENKNDMVEMDIVEKLVKMIPCEHEDLLNITLRLLLNLSFDTGLRNKMVQVGLLPKLTALLGNDNYKQIAMCVLYHISMDDRFKSMFAYTDCIPQLMKMLFECSDERIDLELISFCINLAANKRNVQLICEGNGLKMLMKRALKFKDPLLMKMIRNISQHDGPTKNLFIDYVGDLAAQISNDEEEEFVIECLGTLANLTIPDLDWELVLKEYKLVPYLKDKLKPGAAEDDLVLEVVIMIGTVSMDDSCAALLAKSGIIPALIELLNAQQEDDEFVCQIIYVFYQMVFHQATRDVIIKETQAPAYLIDLMHDKNNEIRKVCDNTLDIIAEYDEEWAKKIQSEKFRWHNSQWLEMVESRQMDESEQYLYGDDRIEPYIHEGDILERPDLFYNSDGLIASEGAISPDFFNDYHLQNGDVVGQHSFPGSLGMDGFGQPVGILGRPATAYGFRPDEPYYYGYGS
Specifications

Specifications

ABM85040.1
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
113.52kDa
Glutathione Sepharose 4 Fast Flow
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
FLJ22818/KAP3/SMAP/Smg-GDS/dJ190I16.1
KIFAP3
Recombinant
wheat germ expression system
Antibody Production, Protein Array, ELISA, Western Blot
22920
KIFAP3 (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue
MQGEDARYLKRKVKGGNIDVHPSEKALIVHYEVEATILGEMGDPMLGERKECQKIIRLKSLNANTDITSLARKVVEECKLIHPSKLNEVEQLLYYLQNRRDSLSGKEKKEKSSKPKDPPPFEGMEIDEVANINDMDEYIELLYEDIPDKVRGSALILQLARNPDNLEELLLNETALGALARVLREDWKQSVELATNIIYIFFCFSSFSQFHGLITHYKIGALCMNIIDHELKRHELWQEELSKKKKAVDEDPENQTLRKDYEKTFKKYQGLVVKQEQLLRVALYLLLNLAEDTRTELKMRNKNIVHMLVKALDRDNFELLILVVSFLKKLSIFMENKNDMVEMDIVEKLVKMIPCEHEDLLNITLRLLLNLSFDTGLRNKMVQVGLLPKLTALLGNDNYKQIAMCVLYHISMDDRFKSMFAYTDCIPQLMKMLFECSDERIDLELISFCINLAANKRNVQLICEGNGLKMLMKRALKFKDPLLMKMIRNISQHDGPTKNLFIDYVGDLAAQISNDEEEEFVIECLGTLANLTIPDLDWELVLKEYKLVPYLKDKLKPGAAEDDLVLEVVIMIGTVSMDDSCAALLAKSGIIPALIELLNAQQEDDEFVCQIIYVFYQMVFHQATRDVIIKETQAPAYLIDLMHDKNNEIRKVCDNTLDIIAEYDEEWAKKIQSEKFRWHNSQWLEMVESRQMDESEQYLYGDDRIEPYIHEGDILERPDLFYNSDGLIASEGAISPDFFNDYHLQNGDVVGQHSFPGSLGMDGFGQPVGILGRPATAYGFRPDEPYYYGYGS
RUO
KIFAP3
Wheat Germ (in vitro)
GST
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers