Learn More
Human Jak3 Antibody, R&D Systems™
GREENER_CHOICE
Mouse Monoclonal Antibody
Brand:  R&D Systems MAB46991-SP
Product Code. 18712939
  • ONLINE EXCLUSIVE PRICE
    201.00€
    190.05€ / 25µg
    Save 10.95€
    5% Off
Description
Jak3 Monoclonal specifically detects Jak3 in Human samples. It is validated for Intracellular Staining by Flow Cytometry, Immunocytochemistry, CyTOF-ready.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Jak3
Monoclonal
LYOPH
Intracellular Staining by Flow Cytometry 2.5 ug/10^6 cells, Immunocytochemistry 8-25 ug/mL, CyTOF-ready
P52333
JAK3
E. coli-derived recombinant human Jak3 Gly46-Thr209 Accession # P52333
25 μg
Primary
Detects human Jak3 in direct ELISAs.
Human
Purified
Flow Cytometry, Immunocytochemistry, CyTOF
452524
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
EC 2.7.10, JAK3_HUMAN, JAK-3EC 2.7.10.2, JAKL, JAKLtyrosine-protein kinase JAK3, Janus kinase 3Leukocyte janus kinase, LJAK, L-JAKJanus kinase 3 (a protein tyrosine kinase, leukocyte)
Mouse
Protein A or G purified from hybridoma culture supernatant
RUO
3718
Reconstitute at 0.5 mg/mL in sterile PBS.
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
IgG2b