missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human ITR Partial ORF (NP_851320.1, 21 a.a. - 120 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00160897-Q01.10ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16192237

  • 349.00€ / 10µg

Estimated Shipment: 08-11-2023
to see stock.
Add to basket

Description

Description

This gene encodes a protein that is a member of the G protein-coupled receptor superfamily. This protein is produced predominantly in vascular smooth muscle cells and may play an important role in the regulation of vascular remodeling. [provided by RefSeq]

Sequence: QGKTLRGSFSSTAAQDAQGQRIGHFEFHGDHALLCVRINNIAVAVGKEAKLYLFQAQEWLKLQQSSHGYSCSEKLSKAQLTMTMNQTEHNLTVSQIPSPQ
Specifications

Specifications

NP_851320.1
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
36.74kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
QGKTLRGSFSSTAAQDAQGQRIGHFEFHGDHALLCVRINNIAVAVGKEAKLYLFQAQEWLKLQQSSHGYSCSEKLSKAQLTMTMNQTEHNLTVSQIPSPQ
RUO
GPR180
Wheat Germ (in vitro)
GST
Liquid
Antibody Production, ELISA, Protein Array, Western Blot
160897
ITR (Human) Recombinant Protein (Q01)
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
ITR
GPR180
Recombinant
wheat germ expression system
Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers