missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human ITR Partial ORF (NP_851320.1, 21 a.a. - 120 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
335.00€ - 508.00€
Specifications
Accession Number | NP_851320.1 |
---|---|
For Use With (Application) | Antibody Production, ELISA, Protein Array, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 160897 |
Molecular Weight (g/mol) | 36.74kDa |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16102247
|
Abnova™
H00160897-Q01.25UG |
25 ug |
508.00€
25µg |
Estimated Shipment: 10-05-2024
Log in to see stock. |
|||||
16192237
|
Abnova™
H00160897-Q01.10UG |
10 ug |
335.00€
10µg |
Estimated Shipment: 10-05-2024
Log in to see stock. |
|||||
Description
This gene encodes a protein that is a member of the G protein-coupled receptor superfamily. This protein is produced predominantly in vascular smooth muscle cells and may play an important role in the regulation of vascular remodeling. [provided by RefSeq]
Sequence: QGKTLRGSFSSTAAQDAQGQRIGHFEFHGDHALLCVRINNIAVAVGKEAKLYLFQAQEWLKLQQSSHGYSCSEKLSKAQLTMTMNQTEHNLTVSQIPSPQSpecifications
NP_851320.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.74kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
ITR | |
GPR180 | |
Recombinant | |
wheat germ expression system |
Antibody Production, ELISA, Protein Array, Western Blot | |
160897 | |
ITR (Human) Recombinant Protein (Q01) | |
QGKTLRGSFSSTAAQDAQGQRIGHFEFHGDHALLCVRINNIAVAVGKEAKLYLFQAQEWLKLQQSSHGYSCSEKLSKAQLTMTMNQTEHNLTVSQIPSPQ | |
RUO | |
GPR180 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |