missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Irisin/FNDC5 Antibody, R&D Systems™

Mouse Monoclonal Antibody
210.00€ - 589.00€
Specifications
| Antigen | Irisin/FNDC5 |
|---|---|
| Clone | 1056126 |
| Dilution | Immunohistochemistry 5-25 ug/mL |
| Applications | Immunohistochemistry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18773497
![]() |
R&D Systems
MAB94202-SP |
25 μg |
210.00€
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18742057
![]() |
R&D Systems
MAB94202-100 |
100 μg |
589.00€
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Irisin/FNDC5 Monoclonal antibody specifically detects Irisin/FNDC5 in Human samples. It is validated for ImmunohistochemistrySpecifications
| Irisin/FNDC5 | |
| Immunohistochemistry 5-25 ug/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied either lyophilized or as a 0.2 μm filtered solution in PBS. | |
| fibronectin type III domain containing 5, fibronectin type III domain-containing protein 5, FNDC5, FRCP2, FRCP2Fibronectin type III repeat-containing protein 2 | |
| Synthetic peptide covering the C-term of the Irisin peptide sequences of the FNDC5 gene., Glu97-Glu143, Accession # Q8NAU1 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. |
| 1056126 | |
| Immunohistochemistry | |
| Unconjugated | |
| Mouse | |
| Human | |
| Q8NAU1 | |
| 252995 | |
| IgG2a | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20 to -70 °C under sterile conditions after reconstitution. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title