missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human IL1F6 Partial ORF (NP_055255.1, 1 a.a. - 61 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00027179-Q01.10ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16195766

  • 349.00€ / 10µg

Estimated Shipment: 08-11-2023
to see stock.
Add to basket

Description

Description

Sequence: MEKALKIDTPQRGSIQDINHRVWVLQDQTLIAVPRKDRMSPVTIALISCRHVETLEKDRGN
Specifications

Specifications

NP_055255.1
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
32.45kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
MEKALKIDTPQRGSIQDINHRVWVLQDQTLIAVPRKDRMSPVTIALISCRHVETLEKDRGN
RUO
IL1F6
Wheat Germ (in vitro)
GST
Liquid
Antibody Production, ELISA, Protein Array, Western Blot
27179
IL1F6 (Human) Recombinant Protein (Q01)
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
FIL1/FIL1(EPSILON)/FIL1E/IL-1F6/IL1(EPSILON)/MGC129552/MGC129553
IL1F6
Recombinant
wheat germ expression system
Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers