missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human IL1F6 Partial ORF (NP_055255.1, 1 a.a. - 61 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
335.00€ - 508.00€
Specifications
Accession Number | NP_055255.1 |
---|---|
For Use With (Application) | Antibody Production, ELISA, Protein Array, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 27179 |
Molecular Weight (g/mol) | 32.45kDa |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16105776
|
Abnova™
H00027179-Q01.25UG |
25 ug |
508.00€
25µg |
Estimated Shipment: 05-06-2024
Log in to see stock. |
|||||
16195766
|
Abnova™
H00027179-Q01.10UG |
10 ug |
335.00€
10µg |
Estimated Shipment: 05-06-2024
Log in to see stock. |
|||||
Description
Sequence: MEKALKIDTPQRGSIQDINHRVWVLQDQTLIAVPRKDRMSPVTIALISCRHVETLEKDRGNSpecifications
NP_055255.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
32.45kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
FIL1/FIL1(EPSILON)/FIL1E/IL-1F6/IL1(EPSILON)/MGC129552/MGC129553 | |
IL1F6 | |
Recombinant | |
wheat germ expression system |
Antibody Production, ELISA, Protein Array, Western Blot | |
27179 | |
IL1F6 (Human) Recombinant Protein (Q01) | |
MEKALKIDTPQRGSIQDINHRVWVLQDQTLIAVPRKDRMSPVTIALISCRHVETLEKDRGN | |
RUO | |
IL1F6 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |