missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Recombinant Human IL-6/IL-6R alpha Protein Chimera

Product Code. 18767626
Click to view available options
Quantity:
25 μg
Unit Size:
25µg
This item is not returnable. View return policy

Product Code. 18767626

Brand: R&D Systems™ 8954SR025

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility.

The Recombinant Human IL-6/IL-6R alpha Protein Chimera is derived from HEK293. The Recombinant Human IL-6/IL-6R alpha Protein Chimera has been validated for the following applications: Bioactivity.
TRUSTED_SUSTAINABILITY

Specifications

For Use With (Application) Bioactivity
Gene ID (Entrez) 3570
Molecular Weight (g/mol) Observed M.W.: 78-93kDa, reducing conditions, Theoretical M.W.: 59kDa
Name IL-6R alpha
Quantity 25 μg
Source Human embryonic kidney cell,HEK293-derived human IL-6/IL-6R alpha Complex protein, (N-terminus) Human IL-6/IL-6R alpha (Leu20-Asp358) Accession &Num; P08887 GGGSGGGSGGGS Human IL-6 Val30-Met212 Accession &Num; P05231 (C-terminus)
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Endotoxin Concentration <0.10 EU per 1μg of the protein by the LAL method.
Gene Alias BSF-2, CD126, gp80, IL6, IL-6 R alpha, IL6Q, IL6R, IL6R alpha, IL-6R-1, IL6RA, IL-6RA, IL6RQ, interleukin 6 receptor, MGI-2A
Common Name IL-6 R alpha
Gene Symbol IL6
Biological Activity Measured by its ability to inhibit M1 mouse myeloid leukemia cell proliferation. The ED50 for this effect is typically 2.5-12.5ng/mL.
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Expression System Human embryonic kidney cell, HEK293-derived Human IL-6 R alpha (Leu20-Asp358) Accession # P08887, N-terminus; GGGSGGGSGGGS; Human IL-6 Val30-Met212 Accession # P05231, C-terminus
Purity or Quality Grade >90%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.