missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Interleukin 2 Recombinant Protein, Invitrogen™
Click to view available options
Quantity:
1 Vial
Conditionnement:
Each
Description
Interleukin 2 Recombinant Protein, SIL2, from Invitrogen.
Host Species: E. Coli
Formulation: 1 Vial purified protein (3.75 ng/ml) with 4% BSA and no preservative
The human IL-2 Standard is calibrated against NIBSC IL-2 Reference Standard lot #86/504. 1 Thermo pg = 0.77 NIBSC pg = 0.014 NIBSC units. Lyophilized product is shipped on cold packs. Stable until expiration if stored desiccated at 2-8°C. Reconstituted cytokine standards should be stored in aliquots at -70°C. Each aliquot should be used only once and discarded afterwards. Stability of reconstituted and frozen cytokine standards may vary. Controls should be run with each standard curve. Sufficient For: ELISA standard curve with replicates or as a control in Western blots. 3750 pg/ml when reconstituted at the recommend volume. Reconstitute with distilled water at volume indicated on label. Use or freeze product within 1 hour of reconstitution.
Spécification
Spécification
| For Use With (Application) | ELISA, Western Blot |
| Formulation | 1 Vial purified protein (3.75 ng/ml) with 4% BSA and no preservative |
| Quantity | 1 Vial |
| Conjugate | Unconjugated |
| Species | E. coli |
| Form | Purified |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu