missing translation for 'onlineSavingsMsg'
Learn More

Human IL-17F Antibody, R&D Systems™
GREENER_CHOICE

Mouse Monoclonal Antibody

238.00€ - 516.00€

Specifications

Antigen IL-17F
Clone 775602R
Dilution ELISA
Applications ELISA
Classification Monoclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18749617
GREENER_CHOICE
View Documents
R&D Systems
MAB13353-SP
25 μg
238.00€
25µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18784819
GREENER_CHOICE
View Documents
R&D Systems
MAB13353-100
100 μg
516.00€
100µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

IL-17F Monoclonal specifically detects IL-17F in Human samples. It is validated for ELISA.
Specifications

Specifications

IL-17F
ELISA
Monoclonal
Supernatant
Human
Q96PD4
112744
IgG2b
Protein A or G purified from cell culture supernatant
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
775602R
ELISA
Unconjugated
Mouse
Lyophilized from a 0.2μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2μm filtered solution in PBS.
Cytokine ML-1, IL17F, IL-17Finterleukin-17F, interleukin 17F
Mouse myeloma cell line NS0-derived recombinant human IL-17F Gly21-Thr153 Accession # Q96PD4
Primary
Reconstitute at 0.5mg/mL in sterile PBS.
Detects human IL-17F in direct ELISAs.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.