missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Interleukin 16 Recombinant Protein, Invitrogen™
Click to view available options
Quantity:
1 Vial
Packungsgröße:
Each
Beschreibung
Interleukin 16 Recombinant Protein, SIL16, from Invitrogen.
Host Species: E. Coli
Formulation: 1 Vial purified protein (5 ng/ml) with 4% BSA and no preservative
International reference standard is not available. Calibrated to a Thermo reference standard. Lyophilized product is shipped on cold packs. Stable until expiration if stored desiccated at -20°C. Reconstituted cytokine standards should be stored in aliquots at -70°C. Each aliquot should be used only once and discarded afterwards. Stability of reconstituted and frozen cytokine standards may vary. Controls should be run with each standard curve. Sufficient For: ELISA standard curve with replicates or as a control in Western blots. 5000 pg/ml when reconstituted at the recommend volume Reconstitute with distilled water at volume indicated on label. Use or freeze product within 1 hour of reconstitution.
Spezifikation
Spezifikation
| For Use With (Application) | ELISA, Western Blot |
| Formulation | 1 Vial purified protein (5 ng/ml) with 4% BSA and no preservative |
| Quantity | 1 Vial |
| Conjugate | Unconjugated |
| Species | E. coli |
| Form | Purified |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur