missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human IL-12 R beta 2 Biotinylated Antibody, R&D Systems™
Goat Polyclonal Antibody
Brand: R&D Systems BAF1959
This item is not returnable.
View return policy
Description
IL-12 R beta 2 Polyclonal specifically detects IL-12 R beta 2 in Human samples. It is validated for Western Blot.
Specifications
| IL-12 R beta 2 | |
| Polyclonal | |
| Biotin | |
| Q99665 | |
| IL12RB2 | |
| Mouse myeloma cell line NS0-derived recombinant human IL-12 R beta 2 Met1-Asn622 Accession # Q99665 | |
| 50 μg | |
| Primary | |
| Detects human IL-12 R beta 2 in Western blots. In Western blots, less than 1% cross-reactivity with recombinant human IL-12 R beta 1. | |
| Human | |
| IgG |
| Western Blot | |
| LYOPH | |
| Western Blot 0.1 ug/mL | |
| IL-12 receptor beta 2, IL-12 receptor subunit beta-2, IL12R beta 2, IL-12R subunit beta-2, IL12RB2, IL-12RB2, IL-12R-beta-2, interleukin 12 receptor, beta 2, interleukin-12 receptor beta-2 chain, interleukin-12 receptor subunit beta-2 | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 3595 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction