missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human IL-10 Antibody, R&D Systems™

Rat Monoclonal Antibody
194.00€ - 489.00€
Specifications
| Antigen | IL-10 |
|---|---|
| Clone | 997935 |
| Dilution | Immunohistochemistry 0.5-25 μg;/mL |
| Applications | Immunohistochemistry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18742187
![]() |
R&D Systems
MAB92101-SP |
25 μg |
194.00€
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18703717
![]() |
R&D Systems
MAB92101-100 |
100 μg |
489.00€
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
IL-10 Monoclonal specifically detects IL-10 in Human samples. It is validated for Immunohistochemistry.Specifications
| IL-10 | |
| Immunohistochemistry 0.5-25 μg;/mL | |
| Monoclonal | |
| Supernatant | |
| Human | |
| P22301 | |
| 3586.0 | |
| IgG1 | |
| Protein A or G purified from cell culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution. |
| 997935 | |
| Immunohistochemistry | |
| Unconjugated | |
| Rat | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
| CSIF, CSIFMGC126450, Cytokine synthesis inhibitory factor, GVHDS, IL10, IL10A, IL-10MGC126451, interleukin 10, interleukin-10, TGIF | |
| Human embryonic kidney cell, HEK293-derived human IL-10, Met1-Asn178, Accession No. P22301 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Detects human IL-10 in direct ELISAs. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title