missing translation for 'onlineSavingsMsg'
Learn More

Human IgG APC-conjugated Antibody, R&D Systems™

Product Code. 18758406
Change view
Click to view available options
Quantity:
100 Tests
Unit Size:
100 tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18758406 100 Tests 100 tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18758406 Supplier R&D Systems Supplier No. F0156100
  • ONLINE EXCLUSIVE PRICE
    389.00€
    367.50€ / 100 tests
    Save 21.50€
    5% Off
 Restricted Item
Estimated Shipment: 21-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Rabbit Monoclonal Antibody

IgG Monoclonal specifically detects IgG in Human samples. It is validated for Flow Cytometry.
TRUSTED_SUSTAINABILITY

Specifications

Antigen IgG
Applications Flow Cytometry
Classification Monoclonal
Clone 1268C
Conjugate APC
Dilution Flow Cytometry
Formulation Supplied in a saline solution containing BSA and sodium azide
Gene Alias Immunoglobulin G, ImmunoglobulinG
Host Species Rabbit
Immunogen Purified IgG from human serum
Quantity 100 Tests
Primary or Secondary Secondary
Gene ID (Entrez) 3500
Test Specificity Detects human IgG in direct ELISAs.
Target Species Human
Content And Storage Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8°C as supplied.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.